Assay Details
Gene Symbol: Ctsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022326.3 | NP_071721.2 | 1 - 2 | 95 | 102 | |
GenBank mRNA | AK005428.1 | - | 1 - 2 | 95 | 102 | |
AK005550.1 | - | 1 - 2 | 90 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK005428.1, AK005550.1, NM_022326.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
