Assay Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023290.2 | NP_075779.2 | 3 - 4 | 394 | 110 | |
| GenBank mRNA | AF277171.1 | - | 3 - 4 | 362 | 110 | |
| AK009515.1 | - | 3 - 4 | 403 | 110 | ||
| AK011295.1 | - | 3 - 4 | 373 | 110 | ||
| AK077926.1 | - | 3 - 4 | 394 | 110 | ||
| BC025547.1 | - | 3 - 4 | 413 | 5320887 | 110 | |
More Information
Set Membership: |
|

Mkrn2
Genomic Map