Assay Details
| Gene Symbol: U2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163769.1 | NP_001157241.1 | 1 - 2 | 103 | 126 | |
| NM_024187.4 | NP_077149.2 | 1 - 2 | 103 | 126 | ||
| GenBank mRNA | AK008332.1 | - | 1 - 2 | 65 | 126 | |
| AK012334.1 | - | 1 - 2 | 63 | 126 | ||
| AK012849.1 | - | 1 - 2 | 63 | 126 | ||
| AK088979.1 | - | 1 - 2 | 73 | 126 | ||
| AK150872.1 | - | 1 - 2 | 103 | 126 | ||
| AV453783.1 | - | 1 - 2 | 85 | 126 | ||
| BC115479.1 | - | 1 - 2 | 65 | 40100558 | 126 | |
| BC115480.1 | - | 1 - 2 | 65 | 40100559 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008332.1, AK012849.1, AK088979.1, AK150872.1, BC115479.1, BC115480.1, NM_001163769.1, NM_024187.4 transcripts.
Set Membership: |
|

U2af1
Genomic Map