Assay Details
| Gene Symbol: Oas1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145211.2 | NP_660212.2 | 2 - 3 | 519 | 90 | |
| GenBank mRNA | AK149613.1 | - | 2 - 3 | 480 | 90 | |
| BC013715.1 | - | 2 - 3 | 501 | 3257027 | 90 | |
| BC057878.1 | - | 2 - 3 | 478 | 5348858 | 90 | |
| M33863.1 | - | 505 | 90 | |||
| X04958.1 | - | 2 - 3 | 505 | 90 | ||
More Information
Set Membership: |
|

Oas1a
Genomic Map