Assay Details
| Gene Symbol: Rdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147222.2 | NP_671755.2 | 2 - 3 | 669 | 114 | |
| GenBank mRNA | AY046408.1 | - | 2 - 3 | 665 | 114 | |
More Information
Set Membership: |
|

Rdh19
Genomic Map