Assay Details
| Gene Symbol: Pnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123371.2 | NP_001116843.1 | 6 | 1014 | 94 | |
| GenBank mRNA | AK008143.1 | - | 6 | 1018 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Pnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123371.2 | NP_001116843.1 | 6 | 1014 | 94 | |
| GenBank mRNA | AK008143.1 | - | 6 | 1018 | 94 | |
Set Membership: |
|