Assay Details
| Gene Symbol: Usp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137547.1 | NP_001131019.1 | 2 | 1726 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp51| Gene Symbol: Usp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137547.1 | NP_001131019.1 | 2 | 1726 | 84 | |
Set Membership: |
|