Assay Details
| Gene Symbol: Fam84b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162926.1 | NP_001156398.1 | ||||
| GenBank mRNA | AK052388.1 | - | ||||
| AK139928.1 | - | |||||
| AK162412.1 | - | |||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam84b| Gene Symbol: Fam84b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162926.1 | NP_001156398.1 | ||||
| GenBank mRNA | AK052388.1 | - | ||||
| AK139928.1 | - | |||||
| AK162412.1 | - | |||||