Assay Details
| Gene Symbol: Ccl27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164044.1 | NP_001157516.1 | 4 | 616 | 85 | |
| NM_001164045.1 | NP_001157517.1 | 3 | 837 | 85 | ||
More Information
Set Membership: |
|

Ccl27a
Genomic Map