Assay Details
| Gene Symbol: Emc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168470.1 | NP_001161942.1 | 2 | 417 | 81 | |
| NM_025318.3 | NP_079594.1 | 2 | 368 | 81 | ||
| GenBank mRNA | AK002385.1 | - | 2 | 353 | 81 | |
| AK003198.1 | - | 2 | 363 | 81 | ||
| AK004149.1 | - | 2 | 364 | 81 | ||
| AK012769.1 | - | 2 | 353 | 81 | ||
| BC022104.1 | - | 2 | 339 | 5101119 | 81 | |
More Information
Set Membership: |
|

Emc6
Genomic Map