Assay Details
| Gene Symbol: Zfp708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012325.2 | NP_001012325.2 | 4 | 2250 | 115 | |
| NM_001012448.2 | NP_001012450.2 | 4 | 2162 | 115 | ||
| NM_001012449.2 | NP_001012451.2 | 3 | 2154 | 115 | ||
| NM_001292066.1 | NP_001278995.1 | 4 | 2119 | 115 | ||
| GenBank mRNA | AK132353.1 | - | 4 | 2124 | 115 | |
| BC089368.1 | - | 4 | 2129 | 115 | ||
More Information
Set Membership: |
|

Zfp708
Genomic Map