Assay Details
Gene Symbol: Defb36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037247.4 | NP_001032324.1 | 2 | 422 | 75 | |
GenBank mRNA | AK005872.1 | - | 2 | 377 | 75 | |
BX631384.1 | - | 1 | 49 | 75 | ||
DQ012042.1 | - | 2 | 422 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
