Assay Details
| Gene Symbol: Zfp950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047931.1 | - | 1 | 731 | 108 | |
| BC029127.1 | - | 2 | 1457 | 108 | ||
| BC040780.1 | - | 2 | 1457 | 108 | ||
More Information
Set Membership: |
|

Zfp950
Genomic Map