Assay Details
| Gene Symbol: Art2a-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007490.1 | NP_031516.1 | 513 | 125 | ||
| NR_033804.1 | 3 | 583 | 125 | |||
| GenBank mRNA | AF016462.1 | - | 513 | 125 | ||
| AF016464.1 | - | 2 | 513 | 125 | ||
| AK018555.1 | - | 3 | 584 | 125 | ||
| BC120753.1 | - | 607 | 40129676 | 125 | ||
| S79913.1 | - | 525 | 125 | |||
| X52991.1 | - | 513 | 125 | |||
More Information
Set Membership: |
|

Art2a-ps
Genomic Map