Assay Details
| Gene Symbol: Hmgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008252.3 | NP_032278.1 | 5 | 2425 | 68 | |
| GenBank mRNA | AK135296.1 | - | 5 | 2330 | 68 | |
| AK135297.1 | - | 5 | 2330 | 68 | ||
More Information
Set Membership: |
|

Hmgb2
Genomic Map