Assay Details
| Gene Symbol: Sox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009237.2 | NP_033263.2 | 1 | 724 | 148 | |
| GenBank mRNA | AB108674.1 | - | 1 | 499 | 148 | |
| BC052024.1 | - | 1 | 721 | 148 | ||
| BC063061.1 | - | 1 | 718 | 148 | ||
| BC096018.1 | - | 1 | 682 | 148 | ||
More Information
Set Membership: |
|

Sox3
Genomic Map