Assay Details
| Gene Symbol: H2-DMb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010388.4 | NP_034518.1 | 2 | 216 | 109 | |
| GenBank mRNA | AK154923.1 | - | 2 | 320 | 109 | |
| BC052864.1 | - | 3 | 471 | 109 | ||
| BC141105.1 | - | 2 | 282 | 9055717 | 109 | |
| BC145385.1 | - | 2 | 282 | 9053943 | 109 | |
| BC145386.1 | - | 2 | 282 | 9053944 | 109 | |
| BC171960.1 | - | 2 | 282 | 9054246 | 109 | |
| KJ650224.1 | - | 2 | 216 | 109 | ||
| U35334.1 | - | 2 | 216 | 109 | ||
| U35335.1 | - | 2 | 216 | 109 | ||
| U35336.1 | - | 2 | 216 | 109 | ||
| U35337.1 | - | 2 | 216 | 109 | ||
| U35338.1 | - | 2 | 216 | 109 | ||
| X90796.1 | - | 2 | 216 | 109 | ||
| X90797.1 | - | 2 | 216 | 109 | ||
| X90798.1 | - | 2 | 216 | 109 | ||
| X90799.1 | - | 2 | 216 | 109 | ||
| X90800.1 | - | 2 | 216 | 109 | ||
| X90801.1 | - | 2 | 216 | 109 | ||
| X90802.1 | - | 2 | 216 | 109 | ||
| X90803.1 | - | 2 | 216 | 109 | ||
| X90804.1 | - | 2 | 216 | 109 | ||
| X90805.1 | - | 2 | 216 | 109 | ||
| X90806.1 | - | 2 | 216 | 109 | ||
| X90807.1 | - | 2 | 216 | 109 | ||
| X90808.1 | - | 2 | 216 | 109 | ||
More Information
Set Membership: |
|

H2-DMb2
Genomic Map