Assay Details
| Gene Symbol: Wbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011712.2 | NP_035842.1 | 3 | 318 | 113 | |
| GenBank mRNA | AK002214.1 | - | 3 | 272 | 113 | |
| AK017717.1 | - | 3 | 279 | 113 | ||
| AY459189.1 | - | 3 | 317 | 113 | ||
| BC007478.1 | - | 3 | 215 | 3490650 | 113 | |
| BC020289.1 | - | 3 | 243 | 113 | ||
| BU511227.1 | - | 3 | 349 | 113 | ||
| U92454.1 | - | 1 | 213 | 113 | ||
More Information
Set Membership: |
|

Wbp5
Genomic Map