Assay Details
| Gene Symbol: Toporsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026652.2 | NP_080928.1 | 2 | 2220 | 110 | |
| GenBank mRNA | AK006061.1 | - | 1 | 1730 | 110 | |
| AK019748.1 | - | 2 | 2221 | 110 | ||
| BC120821.1 | - | 1 | 2089 | 40129744 | 110 | |
| BC120823.1 | - | 1 | 2089 | 40129746 | 110 | |
More Information
Set Membership: |
|

Toporsl
Genomic Map