Assay Details
| Gene Symbol: Wdr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027149.2 | NP_081425.2 | 3 | 1867 | 132 | |
| GenBank mRNA | AK052481.1 | - | 3 | 1814 | 132 | |
| AK136417.1 | - | 3 | 1827 | 132 | ||
| AK145389.1 | - | 3 | 1790 | 132 | ||
| BC026761.1 | - | 4 | 1717 | 4221717 | 132 | |
| BC064773.1 | - | 3 | 1824 | 132 | ||
More Information
Set Membership: |
|

Wdr20
Genomic Map