Assay Details
| Gene Symbol: Ugt1a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201410.3 | NP_958812.3 | 2 | 423 | 98 | |
| GenBank mRNA | AY227198.1 | - | 1 | 379 | 98 | |
| BC026561.1 | - | 2 | 403 | 5050488 | 98 | |
More Information
Set Membership: |
|

Ugt1a6b
Genomic Map