Assay Details
| Gene Symbol: Il5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010558.1 | NP_034688.1 | 1 - 2 | 182 | 84 | |
| GenBank mRNA | BC125366.1 | - | 1 - 2 | 167 | 40129881 | 84 |
| BC132066.1 | - | 1 - 2 | 167 | 40130343 | 84 | |
| X04601.1 | - | 1 - 2 | 182 | 84 | ||
| X06270.1 | - | 1 - 2 | 182 | 84 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Citations (1)
Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
- Authors
- Chen G, Wan H, Luo F, Zhang L, Xu Y, Lewkowich I, Wills-Karp M, Whitsett JA
- Journal
- J Immunol - Page No 6133 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 184 (2010)
- Product Usage
- -

Il5
Genomic Map