Assay Details
| Gene Symbol: Ccl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009139.3 | NP_033165.1 | 1 - 2 | 219 | 99 | |
| GenBank mRNA | AF128188.1 | - | 1 - 2 | 115 | 99 | |
| AF128189.1 | - | 1 - 2 | 115 | 99 | ||
| AF128190.1 | - | 1 - 2 | 115 | 99 | ||
| AF128191.1 | - | 1 - 2 | 115 | 99 | ||
| AF128192.1 | - | 1 - 2 | 115 | 99 | ||
| AK002697.1 | - | 1 - 2 | 122 | 99 | ||
| AK008547.1 | - | 1 - 2 | 123 | 99 | ||
| AK150336.1 | - | 1 - 2 | 122 | 99 | ||
| AK154553.1 | - | 1 - 2 | 123 | 99 | ||
| BC002073.1 | - | 1 - 2 | 112 | 3492808 | 99 | |
| BY172106.1 | - | 1 - 2 | 219 | 99 | ||
| M58004.1 | - | 1 - 2 | 122 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl6
Genomic Map