Assay Details
| Gene Symbol: Ccr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009912.4 | NP_034042.3 | 2 | 506 | 145 | |
| GenBank mRNA | AK031109.1 | - | 2 | 507 | 145 | |
| AK036597.1 | - | 2 | 208 | 145 | ||
| AK036690.1 | - | 2 | 506 | 145 | ||
| AK076275.1 | - | 2 | 381 | 145 | ||
| BC011092.1 | - | 2 | 209 | 3992755 | 145 | |
| CT010178.1 | - | 1 | 41 | 145 | ||
| U29678.1 | - | 1 | 41 | 145 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccr1
Genomic Map