Assay Details
| Gene Symbol: Cxcr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007551.2 | NP_031577.2 | 1 - 2 | 98 | 74 | |
| GenBank mRNA | AK089611.1 | - | 1 - 2 | 111 | 74 | |
| AK133064.1 | - | 3 - 4 | 330 | 74 | ||
| AK155954.1 | - | 1 - 2 | 95 | 74 | ||
| AK156087.1 | - | 1 - 2 | 95 | 74 | ||
| AK157016.1 | - | 1 - 2 | 95 | 74 | ||
| BC064059.1 | - | 1 - 2 | 98 | 1366773 | 74 | |
| X71788.1 | - | 1 - 2 | 55 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cxcr5
Genomic Map