Assay Details
| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021283.2 | NP_067258.1 | 2 - 3 | 238 | 68 | |
| GenBank mRNA | AF352783.1 | - | 2 - 3 | 219 | 68 | |
| BC027514.1 | - | 2 - 3 | 217 | 3376587 | 68 | |
| M13238.1 | - | 2 - 3 | 234 | 68 | ||
| M25892.1 | - | 2 - 3 | 266 | 68 | ||
| X03532.1 | - | 2 - 3 | 240 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il4
Genomic Map