Assay Details
| Gene Symbol: NR2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162543.1 | NP_001156015.1 | 2 - 3 | 569 | 66 | |
| XM_027956971.1 | XP_027812772.1 | 2 - 3 | 1756 | 66 | ||
| GenBank mRNA | FJ969420.1 | - | 2 - 3 | 569 | 66 | |
More Information
Set Membership: |
|

NR2F2
Genomic Map