Assay Details
| Gene Symbol: LCORL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970889.1 | XP_027826690.1 | 3 - 4 | 300 | 105 | |
| XM_027970890.1 | XP_027826691.1 | 3 - 4 | 300 | 105 | ||
| XM_027970891.1 | XP_027826692.1 | 3 - 4 | 300 | 105 | ||
| XM_027970892.1 | XP_027826693.1 | 3 - 4 | 300 | 105 | ||
| XM_027970893.1 | XP_027826694.1 | 3 - 4 | 300 | 105 | ||
| XM_027970894.1 | XP_027826695.1 | 3 - 4 | 300 | 105 | ||
More Information
Set Membership: |
|

LCORL
Genomic Map