Assay Details
| Gene Symbol: PKNOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959267.1 | XP_027815068.1 | 10 - 11 | 1303 | 66 | |
| XM_027959268.1 | XP_027815069.1 | 10 - 11 | 1233 | 66 | ||
More Information
Set Membership: |
|

PKNOX2
Genomic Map