Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977051.1 | XP_027832852.1 | 2 - 3 | 357 | 74 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PEA15| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977051.1 | XP_027832852.1 | 2 - 3 | 357 | 74 | |