Assay Details
| Gene Symbol: ARL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126367.3 | NP_001119839.2 | 4 - 5 | 397 | 88 | |
| XM_027965716.1 | XP_027821517.1 | 3 - 4 | 394 | 88 | ||
| GenBank mRNA | EU596599.1 | - | 3 - 4 | 331 | 88 | |
| GT644091.1 | - | 4 - 5 | 429 | 88 | ||
More Information
Set Membership: |
|

ARL1
Genomic Map