Assay Details
| Gene Symbol: WASF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011209.4 | XP_004011258.1 | 6 - 7 | 1001 | 79 | |
| XM_012182636.3 | XP_012038026.1 | 6 - 7 | 996 | 79 | ||
| XM_027972375.1 | XP_027828176.1 | 6 - 7 | 1026 | 79 | ||
| XM_027972376.1 | XP_027828177.1 | 6 - 7 | 1023 | 79 | ||
More Information
Set Membership: |
|

WASF1
Genomic Map