Assay Details
| Gene Symbol: YTHDF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965411.1 | XP_027821212.1 | 3 - 4 | 462 | 89 | |
| XM_027965412.1 | XP_027821213.1 | 3 - 4 | 445 | 89 | ||
More Information
Set Membership: |
|

YTHDF2
Genomic Map