Assay Details
| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002627.4 | XP_004002676.1 | 3 - 4 | 543 | 76 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRABP2| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002627.4 | XP_004002676.1 | 3 - 4 | 543 | 76 | |