Assay Details
| Gene Symbol: GALNT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012174255.3 | XP_012029645.3 | 6 - 7 | 1425 | 52 | |
| XM_027967307.1 | XP_027823108.1 | 6 - 7 | 1456 | 52 | ||
More Information
Set Membership: |
|

GALNT6
Genomic Map