Assay Details
| Gene Symbol: GSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014515.4 | XP_004014564.2 | 6 - 7 | 692 | 68 | |
| XM_012189114.3 | XP_012044504.2 | 6 - 7 | 633 | 68 | ||
More Information
Set Membership: |
|

GSS
Genomic Map