Assay Details
| Gene Symbol: HDHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103661.3 | XP_011959051.1 | 5 - 6 | 723 | 60 | |
| XM_027960930.1 | XP_027816731.1 | 5 - 6 | 763 | 60 | ||
More Information
Set Membership: |
|

HDHD2
Genomic Map