Assay Details
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005638.4 | XP_004005687.2 | 3 - 4 | 460 | 83 | |
XM_027966446.1 | XP_027822247.1 | 3 - 4 | 289 | 83 | ||
XM_027966447.1 | XP_027822248.1 | 3 - 4 | 269 | 83 | ||
XM_027966448.1 | XP_027822249.1 | 3 - 4 | 269 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
