Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017472.4 | XP_004017521.1 | 3 - 4 | 407 | 76 | |
| XM_012098046.3 | XP_011953436.1 | 3 - 4 | 425 | 76 | ||
More Information
Set Membership: |
|

LIF
Genomic Map