Assay Details
| Gene Symbol: CNGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143663.1 | NP_001137135.1 | 2 - 3 | 216 | 68 | |
| XM_027965727.1 | XP_027821528.1 | 3 - 4 | 428 | 68 | ||
| GenBank mRNA | FM946176.1 | - | 2 - 3 | 216 | 68 | |
| FN377575.1 | - | 2 - 3 | 216 | 68 | ||
| Gene Symbol: LOC114113733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965914.1 | XP_027821715.1 | 2 - 3 | 216 | 68 | |
| GenBank mRNA | FN377574.1 | - | 2 - 3 | 216 | 68 | |
More Information
Set Membership: |
|

CNGA3
Genomic Map