Assay Details
| Gene Symbol: NDUFA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145188.1 | NP_001138660.1 | 3 - 4 | 278 | 70 | |
| GenBank mRNA | FJ546078.1 | - | 3 - 4 | 278 | 70 | |
More Information
Set Membership: |
|

NDUFA12
Genomic Map