Assay Details
| Gene Symbol: ATP6V0D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189960.3 | XP_012045350.2 | 6 - 7 | 963 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V0D1| Gene Symbol: ATP6V0D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189960.3 | XP_012045350.2 | 6 - 7 | 963 | 55 | |
Set Membership: |
|