Assay Details
| Gene Symbol: LOC105608566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096873.2 | XP_014952359.2 | 3 - 4 | 551 | 67 | |
| XM_027971626.1 | XP_027827427.1 | 3 - 4 | 280 | 67 | ||
More Information
Set Membership: |
|

LOC105608566
Genomic Map