Assay Details
| Gene Symbol: SLC2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968628.1 | XP_027824429.1 | 4 - 5 | 711 | 62 | |
| GenBank mRNA | U89029.1 | - | 1 - 2 | 212 | 62 | |
More Information
Set Membership: |
|

SLC2A1
Genomic Map