Assay Details
| Gene Symbol: SLC2A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974995.1 | XP_027830796.1 | 8 - 9 | 1236 | 56 | |
| GenBank mRNA | AY949177.1 | - | 1 - 2 | 76 | 56 | |
More Information
Set Membership: |
|

SLC2A4
Genomic Map