Assay Details
| Gene Symbol: IDH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101962.2 | XP_014957448.2 | 4 - 5 | 334 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDH3A| Gene Symbol: IDH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101962.2 | XP_014957448.2 | 4 - 5 | 334 | 75 | |
Set Membership: |
|