Assay Details
| Gene Symbol: B4GALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246211.1 | NP_001233140.1 | 3 - 4 | 872 | 77 | |
| GenBank mRNA | JF510320.1 | - | 3 - 4 | 872 | 77 | |
More Information
Set Membership: |
|

B4GALT1
Genomic Map