Assay Details
| Gene Symbol: LOC101102072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012749.4 | XP_004012798.1 | 2 - 3 | 792 | 81 | |
| XM_004012750.4 | XP_004012799.1 | 2 - 3 | 406 | 81 | ||
| XM_012185859.2 | XP_012041249.1 | 2 - 3 | 399 | 81 | ||
| XM_027974744.1 | XP_027830545.1 | 2 - 3 | 220 | 81 | ||
More Information
Set Membership: |
|

LOC101102072
Genomic Map