Assay Details
| Gene Symbol: CCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287473.1 | NP_001274402.1 | 3 - 4 | 597 | 72 | |
| XM_004009046.4 | XP_004009095.1 | 4 - 5 | 809 | 72 | ||
| XM_012177580.3 | XP_012032970.1 | 4 - 5 | 805 | 72 | ||
| GenBank mRNA | KC539855.1 | - | 3 - 4 | 597 | 72 | |
More Information
Set Membership: |
|

CCNG1
Genomic Map