Assay Details
| Gene Symbol: PRICKLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962892.1 | XP_027818693.1 | 4 - 5 | 1098 | 72 | |
| XM_027962893.1 | XP_027818694.1 | 5 - 6 | 662 | 72 | ||
| XM_027962894.1 | XP_027818695.1 | 4 - 5 | 1153 | 72 | ||
More Information
Set Membership: |
|

PRICKLE3
Genomic Map